Skip to Content
Merck
All Photos(1)

Key Documents

463922

Sigma-Aldrich

Ammonium metatungstate hydrate

99.99% trace metals basis

Synonym(s):

Ammonium tungsten oxide hydrate, Hexaammonium wolframate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6H2W12O40 · xH2O
CAS Number:
Molecular Weight:
2956.30 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
powder

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

impurities

≤150.0 ppm Trace Metal Analysis

mp

100 °C (dec.) (lit.)

SMILES string

N.N.N.N.N.N.O.O[W]1O[W](O)(=O)(O1)O[W]2(=O)(=O)O[W](O)(=O)(O2)O[W]3(=O)(=O)O[W](O)(=O)(O3)O[W]4(=O)(=O)O[W](O)(=O)(O[W]5(=O)(=O)O[W](O)(=O)(O[W]6(=O)(=O)O[W](O)(O)O6)O5)O4

InChI

1S/6H3N.9H2O.32O.12W/h6*1H3;9*1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*+1;+2/p-8

InChI key

ZBHPUDGBNLMJQH-UHFFFAOYSA-F

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium metatungstate is a Keggin-type heteropoly compound. It can be prepared by polycondensation of ammonium tungstate solutions. It is widely used as a precursor for preparing tungsten catalysts for hydrocracking and as a fuel cell catalyst.

Application

Ammonium metatungstate can be used as a precursor to synthesize:
  • Carbon-supported tungsten catalyst for hydrogenolysis of cellulose.
  • Reduced tungsten and mixed tungsten oxide catalysts for bifunctional hydroisomerization of n-alkanes.
  • Hybrid 2D/3D tungsten diselenide transistors via chemical vapor deposition method.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ze-Da Meng et al.
Nanoscale research letters, 6(1), 459-459 (2011-07-22)
WO3-treated fullerene/TiO2 composites (WO3-fullerene/TiO2) were prepared using a sol-gel method. The composite obtained was characterized by BET surface area measurements, X-ray diffraction, scanning electron microscopy, energy dispersive X-ray analysis, transmission electron microscopy, and UV-vis analysis. A methyl orange (MO) solution
Xiao Zhang et al.
Science advances, 6(40) (2020-10-02)
The direct growth of single-walled carbon nanotubes (SWCNTs) with narrow chiral distribution remains elusive despite substantial benefits in properties and applications. Nanoparticle catalysts are vital for SWCNT and more generally nanomaterial synthesis, but understanding their effect is limited. Solid catalysts
Hanyu Zhang et al.
Nanoscale, 12(15), 8344-8354 (2020-04-03)
By understanding how the environmental composition impacts the optoelectronic properties of transition metal dichalcogenide monolayers, we demonstrate that simple photoluminescence (PL) measurements of tungsten disulfide (WS2) monolayers can differentiate relative humidity environments. In this paper, we examine the PL and
Gwang Hwi An et al.
Small (Weinheim an der Bergstrasse, Germany), 16(43), e2003326-e2003326 (2020-10-01)
While a hexagonal WS2 monolayer, grown by chemical vapor deposition, exhibits distinctive patterns in photoluminescence mapping, segmented with alternating S-vacancy (SV) and W-vacancy (WV) domains in a single crystal, the formation mechanism for native alternating defect domains remains unresolved to
Alessandro Chieregato et al.
ChemSusChem, 8(2), 398-406 (2014-12-10)
Glycerol surplus from biodiesel synthesis still represents a major problem in the biofuel production chain. Meanwhile, those in the acrylic acid market are looking for new processes that are able to offer viable alternatives to propylene-based production. Therefore, acrylic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service