Skip to Content
Merck
All Photos(1)

Key Documents

939358

Sigma-Aldrich

Magnesium acetate tetrahydrate

≥99.9% trace metals basis

Synonym(s):

Magnesium Diacetate Tetrahydrate,, Magnesium diethanoate tetrahydrate, Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein:
3730605
MDL number:
UNSPSC Code:
12352302
Assay:
≥99.9% trace metals basis
98-102% (EDTA, complexometric)
form:
powder or crystals
solid
solubility:
water: soluble

type

(High purity salts)

Quality Level

Assay

≥99.9% trace metals basis
98-102% (EDTA, complexometric)

form

powder or crystals
solid

impurities

<1000 ppm trace metal basis

color

white to off-white

mp

72-75 °C (lit.)
72-75 °C

solubility

water: soluble

density

1.454 g/cm3

anion traces

chloride (Cl-): ≤20 ppm
sulfate (SO42-): <50 ppm

cation traces

Al: <50  ppm
K: <50  ppm
Mg: <100  ppm
Na: <50 ppm
Pb: <50 ppm
Zn: <50 ppm

application(s)

battery manufacturing

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium acetate tetrahydrate is a white hygroscopic compounds and soluble in water. It is the source of Mg2+ ions. It is a source of high purity compounds, catalysts and nanomaterials.

Application

Magnesium acetate tetrahydrate is key components for the synthesis of nanomaterials, nanocrystals, and catalyst due to it solubilty in water. It can be used as
  • A key components for the synthesis of spinel magnesium manganese oxide (Mg0.5Mn2.5O4 ) through one-step colloidal synthesis method. The nanocrystals has exhibited significant electrochemical activities in presence of diverse electrolytes.
  • A starting materials for the production of Mesoporous (ZnO)x(MgO)1−x nanoplates by a template-free solvothermal synthetic method followed by subsequent calcination. These materials exhibit a band gap resulting from the presence of ZnO and MgO. The broad peaks observed in the 400-700 nm range indicate the presence of oxygen vacancy defects on the surface of the (ZnO)x(MgO)1−x nanoplates. These nanocrystals showed superior photocatalytic activities for the degradation of methyl orange (MO) in aqueous solution.
  • To the synthesis of hydrophobic antireflective films of MgF2 with silicon modified with enhenced durability through sol-gel method.
  • As a material for synthesizing carbon nanoribbons using ferrocene at high temperatures. The resulting nanoribbons exhibit a remarkably high surface area and demonstrate a stable reversible capacity of 750 mA h g−1 after 300 cycles in a charge-discharge experiment conducted at 0.5 A g−1. Due to these properties, it would be highly beneficial as an electrode material in electronic devices.

Features and Benefits

Water soluble
Medium purity (99.9%)
Low trace metals in ppm level
Cost effective
Low Chloride and sulfate levels

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous (ZnO)x(MgO)1?x nanoplates: template-free solvothermal synthesis, optical properties, and their applications in water treatment
Lian, J. et al.
Nanoscale, 5(23), 11672-11672 (2013)
Hydrophobic MgF2 antireflective films with enhanced environmental durability by a sol?gel process.
Yan, L et al.
Materials Letters, 129, 156?158-156?158 (2014)
One-pot synthesis of carbon nanoribbons and their enhanced lithium storage performance
Mei, T et al.
Journal of Material Chemistry A, 2(30), 11974?11979-11974?11979 (2014)
Multivalent Electrochemistry of Spinel MgxMn3?xO4 Nanocrystals
Kim, C. et al.
Chemistry and Materials Research, 30(5),, 1496?1504-1496?1504 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service