Skip to Content
Merck
All Photos(2)

Documents

934062

Sigma-Aldrich

Aluminum oxide

greener alternative

alpha phase, nanopowder, 135 nm avg. part. size, 99.9% trace metals basis

Synonym(s):

α-Al2O3, Alpha-alumina, Corundum, Dialuminium trioxide, Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
MDL number:
UNSPSC Code:
12352311
NACRES:
NA.21

Quality Level

Assay

99.9% trace metals basis

form

nanopowder

composition

Al2O3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

avg. part. size

135 nm

particle size

135 nm

mp

2040 °C (lit.)

bulk density

0.18 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum oxide, alpha phase, nanopowder is a high-purity, ultrafine powder with an average particle size of 135 nm. Alpha-phase alumina nanopowder has superior mechanical and thermal stability, high hardness, and wear resistance. Its high dielectric strength and low dielectric loss make it a suitable material for insulators and capacitors in electronic components. Additionally, its biocompatibility, bioactivity, and wear resistance make it an attractive material for biomedical applications. This product has a purity of 99.9%, making it an ideal choice for a range of industrial applications. Our aluminum oxide nanopowder is carefully manufactured to ensure consistent quality and particle size distribution, resulting in superior performance and reliability.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Alpha-phase alumina nanopowder with a purity of 99.9% finds its major applications in the production of advanced ceramics, nanocomposites, and as a catalyst support material. The high surface area and excellent dispersibility of alpha-phase alumina nanoparticles make them ideal as a reinforcing agent in nanocomposites, where they improve the mechanical, thermal, and electrical properties of the composite material. In advanced ceramics, the fine particle size of alpha-phase alumina nanopowder makes it a suitable material for use where a high degree of homogeneity is required. Alpha-phase alumina nanopowder is also used as a support material for catalysts due to its high surface area and excellent thermal stability. It is particularly attractive for catalytic applications due to its low acidity and high chemical inertness.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Al2O3 Coating on Stabilizing LiNi0.4Mn0.4Co0.2O2 Cathodes
Wise A M, et al
Chemistry of Materials, 27, 6146-6154 (2015)
Interface and Material Characterization of Thin ALD-Al2O3 Layers on Crystalline Silicon
Naumann V, et al.
Energy Procedia, 27, 312-318 (2012)
Al2O3-coated LiCoO2 as cathode material for lithium ion batteries
Liu L, et al.
Solid State Ionics, 152-153, 341-346 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service