932027
9,10-Dihydro-9,9-dimethyl-10- (9-phenyl-9H-carbazol-3-yl)-acridine
≥99% (HPLC)
Synonym(s):
P-CzAc
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C33H26N2
CAS Number:
Molecular Weight:
450.57
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.23
Recommended Products
grade
sublimed grade
Quality Level
Assay
≥99% (HPLC)
solubility
chloroform: soluble
dibromomethane: soluble
toluene: soluble
Orbital energy
HOMO 5.7 eV
LUMO 2.5 eV
Related Categories
Application
9,10-Dihydro-9,9-dimethyl-10- (9-phenyl-9H-carbazol-3-yl)-acridine, also known as P-CzAc, is a Hole Transport / Electron Blocking Layer (HTL / EBL) polymer used in organic electronics, such as for solution-processed light emitting diodes (LEDs). Eg: green quantum dot light-emitting diodes (QD-LEDs).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A novel molecular design employing a backbone freezing linker for improved efficiency, sharpened emission and long lifetime in thermally activated delayed fluorescence emitters.
Yu J G, et al.
Journal of Material Chemistry C, 7, 2919-2926 (2019)
Long lifetime blue phosphorescent organic light emitting diodes with an exciton blocking layer.
Seo J A, et al.
Journal of Material Chemistry C, 3, 4640-4645 (2015)
High triplet energy exciplex host derived from a CN modified carbazole based n-type host for improved efficiency and lifetime in blue phosphorescent organic light-emitting diodes.
Shin K S, et al.
Journal of Material Chemistry C, 6, 10308-10314 (2018)
Efficiency enhancement in fluorescent deep-blue OLEDs by boosting singlet exciton generation through triplet fusion and charge recombination rate.
Bae H, et al.
Organic Electronics, 70, 1-6 (2019)
Novel carbazole-acridine-based hole transport polymer for low turn-on voltage of green quantum dot light-emitting diodes.
Kim W C, et al.
Polym. Chem., 12, 4714-4721 (2021)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service