Skip to Content
Merck
All Photos(2)

Key Documents

932051

Sigma-Aldrich

2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine

≥99% (HPLC)

Synonym(s):

26DCzPPy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H27N3
CAS Number:
Molecular Weight:
561.67
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23

grade

sublimed grade

Quality Level

description

μh ≈ 1.0 x 10-5 cm2 V−1 s−1
μe ≈ 1.0 x 10-5 cm2 V−1 s−1

Assay

≥99% (HPLC)

loss

0.5% TGA, > 370 °C (weight loss)

solubility

chloroform: soluble
dichloromethane: soluble
toluene: soluble

fluorescence

λem 410 nm±10 nm in dichloromethane

Orbital energy

HOMO 6.05 eV 
LUMO 2.56 eV 

λ

in dichloromethane

UV absorption

λ: 239 nm±5 nm Amax
λ: 292 nm±5 nm Amax

SMILES string

C1(C2=CC=CC(N3C(C=CC=C4)=C4C5=C3C=CC=C5)=C2)=NC(C6=CC(N7C(C=CC=C8)=C8C9=C7C=CC=C9)=CC=C6)=CC=C1

InChI

1S/C41H27N3/c1-5-22-38-32(16-1)33-17-2-6-23-39(33)43(38)30-14-9-12-28(26-30)36-20-11-21-37(42-36)29-13-10-15-31(27-29)44-40-24-7-3-18-34(40)35-19-4-8-25-41(35)44/h1-27H

InChI key

UFWDOFZYKRDHPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine, also known as 26DCzPPy, is a solution-processable phosphorescent bipolar host material used for highly efficient optoelectronic devices such as OLED displays. It has a μh around 1.0 x 10-5 cm2 V−1 s−1 and a μe of about 1.0 x 10-5 cm2 V−1 s−1.
2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine, can be used in R&D in the field of organic electronics. It is commonly utilized as a host material in the emissive layer of organic light-emitting diodes (OLEDs) devices. provides good film-forming properties, high charge transport efficiency, and an excellent energy level match with various emissive dopants. It can be used as a hole-blocking or electron-blocking layer material in OLED devices. It can be used in organic photovoltaic (OPV) devices as an exciton-blocking layer. It helps in enhancing the charge separation and minimizing exciton recombination, thereby improving the power conversion efficiency of OPV devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-efficiency red, green and blue phosphorescent homojunction organic light-emitting diodes based on bipolar host materials
Cai, Chao, et al.
Organic Electronics, 12, 843-850 (2011)
Interfacial TADF Exciplex as a Tool to Localize Excitons, Improve Efficiency, and Increase OLED Lifetime
Colella, M. et al.
ACS Applied Materials & Interfaces, 10, 40001-40007 (2018)
Pyridine-Containing Bipolar Host Materials for Highly Efficient Blue Phosphorescent OLEDs
Su, Shi-Jia, et al.
Chemistry of Materials, 20, 1691-1693 (2008)
Blue and white phosphorescent organic light emitting diode performance improvement by confining electrons and holes inside double emitting layers
Tsai, Yu-Sheng, et al.
Journal of Luminescence, 153, 312-316 (2014)
Soluble processed low-voltage and high efficiency blue phosphorescent organic light-emitting devices using small molecule host systems
Doh J., et al.
Organic Electronics, 13, 586-592 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service