Skip to Content
Merck
All Photos(3)

Key Documents

929700

Sigma-Aldrich

ElectroGreen®

greener alternative

Xylene substitute for electronics, bio-sourced

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.05
Assay:
≥99% (GC)
grade:
for electronic purposes
form:
liquid

grade

for electronic purposes

Quality Level

vapor pressure

36 mmHg ( 20 °C)

description

RelativeEvaporation rate: 0.89

Hansen SolubilityParameters: SPd = 7.7; SPp = 3.1; SPh = 5.8

Specific Gravity (25°C): 0.8905

Assay

≥99% (GC)

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, Zn, trace (ICP, each)
0.2% water (Karl Fischer)

evapn. residue

≤ 0.05%

color

clear

viscosity

9.3  cP(20 °C)

bp

80-160 °C

suitability

suitable for acidity (<=0.09% as lactic acid)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based(corn/beets-derived) safer alternatives to replace synthetic xylene fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource (verified through C14 ASTM D6866-16testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more details.

This solvent blend consists of n-Butylacetate 50-70%, Ethyl lactate 20-30%, ethanol 20-30%, ethyl acetate 1-5%.

Application

Our bio-sourced Xylene substitute is a solvent blend for electronics applications in both - cleaning and ink formulations. It has a better cleaning power and reduced solvent usage vs. Xylene, and also high performance, excellent solvency, versatile solvent blend for ink formulations with high loading capacity.
It is readily recycled via distillation for repeated reuse, with no Global Warming compounds or Hazardous Air Pollutants.

Storage and Stability

To ensure low water content, handle under inert conditions. Product should be refrigerated upon receipt.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.9 °F

Flash Point(C)

16.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service