Skip to Content
Merck
All Photos(1)

Key Documents

928194

Sigma-Aldrich

Gold nanoparticles

100 nm diameter, azide functionalized, 1ml, OD 50, in water

Synonym(s):

AuNP, GdNP, Gold NP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141717
NACRES:
NA.22

Quality Level

description

Alkyne surface density: 1/nm2

concentration

in water

OD

50

core diam.

100 nm , Coefficient of Variance < 12%

diameter

100 nm

λmax

572 nm

PDI

<0.200

functional group

azide

storage temp.

2-8°C

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azide functionalized gold nanoparticles are suitable for covalent conjugation of any molecule with a terminal alkyne through click chemistry. These gold nanoparticles allow for rapid generation of gold conjugates for subsequent use in downstream assays.

Azide functionalized gold nanoparticles are more than 95% spherical and have uniform size distribution (CV <12%).

Application

Ideal for development of gold conjugates for use in applications such as blotting, lateral flow assays, microscopy, and transmission electron microscopy (TEM).

Storage and Stability

When stored for a long period of time gold nanoparticles may sediment at the bottom of the vial, which is especially prominent for larger particle sizes. Prior to use, re-suspend the sedimented particles by thorough mixing until a homogenous solution is obtained.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bionanoconjugation via click chemistry: The creation of functional hybrids of lipases and gold nanoparticles
Brennan, Jennifer L., et al.
Bioconjugate Chemistry, 17, 1373-1375 (2006)
Chain-like assembly of gold nanoparticles on artificial DNA templates via `click chemistry?
Fischler, Monika, et al.
Chemical Communications (Cambridge, England), 2, 169-171 (2008)
Vivek Poonthiyil et al.
Beilstein journal of organic chemistry, 14, 11-24 (2018-01-31)
Glycoscience, despite its myriad of challenges, promises to unravel the causes of, potential new detection methods for, and novel therapeutic strategies against, many disease states. In the last two decades, glyco-gold nanoparticles have emerged as one of several potential new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service