Skip to Content
Merck
All Photos(1)

Key Documents

926671

Sigma-Aldrich

Upconversion Nanoparticles

Silica coated NaYF4,Yb,Tm@NaYF4,Yb,Nd, 808 excited, blue light

Synonym(s):

UCNPs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

Quality Level

concentration

2 mg/mL in water

matrix active group

OH surface treatment

particle size

50 nm±15 nm

fluorescence

λex 808 nm (blue)
λem 365 nm
λem 450 nm
λem 470 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Applications include:
  • Fluorescent microscopy
  • Deep-tissue bioimaging
  • Nanomedicine
  • Optogenetics
  • Security labelling
  • Volumetric display
Upconversion nanoparticles (ucnps), are the result of a unique optical process in which near-infrared (NIR) light excitation is converted into visable and/or UV emission. Upconversion (UC) utilizes sequential absorption of multiple photons utilizing both lifetime and real ladder-like energy levels of ions in a host lattice to produce a higher energy anti-Stokes luminescence.

This optical features of UCNPs, results in deep tissue penetration and minimal autofluorescence background, for a broad range of applications of UCNP in diagnostics and biomedical imaging systems.

Legal Information

Product of RuixiBiotechCo. Ltd

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shihui Wen et al.
Nature communications, 9(1), 2415-2415 (2018-06-22)
Lanthanide-doped upconversion nanoparticles (UCNPs) are capable of converting near-infra-red excitation into visible and ultraviolet emission. Their unique optical properties have advanced a broad range of applications, such as fluorescent microscopy, deep-tissue bioimaging, nanomedicine, optogenetics, security labelling and volumetric display. However
Daniel J Gargas et al.
Nature nanotechnology, 9(4), 300-305 (2014-03-19)
Imaging at the single-molecule level reveals heterogeneities that are lost in ensemble imaging experiments, but an ongoing challenge is the development of luminescent probes with the photostability, brightness and continuous emission necessary for single-molecule microscopy. Lanthanide-doped upconverting nanoparticles overcome problems
Yujia Liu et al.
Nature, 543(7644), 229-233 (2017-02-23)
Lanthanide-doped glasses and crystals are attractive for laser applications because the metastable energy levels of the trivalent lanthanide ions facilitate the establishment of population inversion and amplified stimulated emission at relatively low pump power. At the nanometre scale, lanthanide-doped upconversion
Upconversion nanoparticles: design, nanochemistry, and applications in theranostics.
Guanying Chen et al.
Chemical reviews, 114(10), 5161-5214 (2014-03-13)
Christoph Drees et al.
Angewandte Chemie (International ed. in English), 55(38), 11668-11672 (2016-08-12)
Upconversion nanoparticles (UCNPs) convert near-infrared into visible light at much lower excitation densities than those used in classic two-photon absorption microscopy. Here, we engineered <50 nm UCNPs for application as efficient lanthanide resonance energy transfer (LRET) donors inside living cells. By

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service