Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Er@NaYF4, Oil soluble core-shell, fluorescence λex 980, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, Oil soluble core shell upconversion nanoparticle, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, Oil soluble core shell,808 excitation, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-NH2 modified core-shell, 980 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4:Yb,Tm@NaYF4, Oil soluble core shell, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Tm@NaYF4,Yb,Nd, 808 excited, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4,Yb,Nd, PEG-COOH modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Tm@NaYF4, 980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4,Yb,Er@NaYF4,Yb,Nd, 808 excited, green
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Er@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Tm@NaYF4, PEG-COOH modified core-shell, fluorescence λex 980, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4-Yb,Tm@NaYF4, PEG-NH2 modified core-shell ucnps,980 excitation, blue light
Sigma-Aldrich
Upconversion Nanoparticles, Silica coated NaYF4-Yb,Er@NaYF4, fluorescence λex 980 nm, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Tm@NaYF4 Yb, Nd, PEG-NH2 modified core-shell, fluorescence λex 808 nm, blue light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-NH2 modified core-shell, 808 excitation, green light
Sigma-Aldrich
Upconversion Nanoparticles, NaYF4,Yb,Er@NaYF4,Yb,Nd, PEG-COOH modified core-shell, 808 excitation, green light