921513
Droplet generation and storage chip
Fluidic 719, COC
Synonym(s):
Microfluidic, Microparticle, Nanoparticle
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
description
Microfludic chip x1
Looking for similar products? Visit Product Comparison Guide
Application
Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (μl to fl) of fluids, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.
Droplet generation and storage chip, Fluidic 719, COC is made of COC (Cyclic olefin copolymer ) and has a storage channel design for optical analysis. The channel design of Fluidic 719 has an additional flow focusing junction and droplet storage in one channel, compared to similar chip designs. The chip contains 2261 storage positions and can be used for a wide-range of applications including droplet-based cell culture/monitoring.
Droplet generation and storage chip, Fluidic 719, COC is made of COC (Cyclic olefin copolymer ) and has a storage channel design for optical analysis. The channel design of Fluidic 719 has an additional flow focusing junction and droplet storage in one channel, compared to similar chip designs. The chip contains 2261 storage positions and can be used for a wide-range of applications including droplet-based cell culture/monitoring.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Recent advances of controlled drug delivery usingmicrofluidic platforms.
Advanced Drug Delivery Reviews, 128, 3-28 (2018)
Microfluidic-assisted fabrication of carriers for controlled drug delivery.
Lab on a chip, 17, 1856-1883 (2017)
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service