B04A-03
CellASIC ONIX plate for bacteria cells (4 chamber)
The B04 plates utilize a microfabricated silicone ceiling with a height similar to bacteria cells to restrict their growth in a single focal plane & maintain x,y position over time.
Synonym(s):
Bacterial culture plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PMMA
glass
polycarbonate
polydimethylsiloxane (PDMS)
manufacturer/tradename
CellASIC® ONIX
W × L × H
85.48 mm × 127.76 mm × 14.35 mm
compatibility
for use with cells sized 0.7–3.5 μm
General description
CellASIC ONIX plate for bacteria cells (4 chambers) is designed microfabricated silicone ceiling with a height similar to bacterial cells to restrict their growth in a single focal plane & maintain x,y position over time.
Application
CellASIC ONIX plate for bacteria cells (4 chambers) has been used in the microfluidic live-cell imaging of bacterial cells.
Legal Information
CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frontiers in microbiology, 11, 417-417 (2020-04-08)
Tuberculosis (TB) results in both morbidity and mortality on a global scale. With drug resistance on the increase, there is an urgent need to develop novel anti-mycobacterials. Thus, we assessed the anti-mycobacterial potency of three novel synthetic peptoids against drug-susceptible
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service