911267
mPEG24-Hydrazide
Synonym(s):
Methoxy-PEG-HZ, mPEG-Hydrazide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
solid
mol wt
average Mn 1200 (by NMR)
reaction suitability
reagent type: chemical modification reagent
reactivity: carboxyl reactive
color
white to off-white
Ω-end
hydrazide
α-end
methoxy
polymer architecture
shape: linear
functionality: monofunctional
storage temp.
−20°C
Application
Polyethylene glycols (PEGs) are FDA approved hydrophilic polymers. They are non-toxic, non-immunogenic and non-antigenic, and are widely used in a varity of biomedical applications such as bioconjugation, drug delivery, surface functionalization, and tissue engineering. Methoxy PEG Hydrazide reacts with aldehydes and ketones to give stable hydrazones in a single step; it can also react with activated carboxylic acids.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service