Skip to Content
Merck
All Photos(1)

Key Documents

911216

Sigma-Aldrich

TU-1

Synonym(s):

4,8-Bis[5-[3-(trifluoromethoxy)phenyl]-2-thienyl]benzo[1,2-c:4,5-c′]bis[1,2,5]thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H12F6N4O2S4
CAS Number:
Molecular Weight:
678.67
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

TU-1: Vapor deposition / coating film forming mobility ~ 1 cm2 / Vs (@ Vsd = 100 V)

Assay

99% (HPLC)

form

powder

color

dark green

storage temp.

2-8°C

Application

TU-1 is a N-type organic semiconductor material that provides both high mobility and high atmospheric stability. It can be dissolved in organic solvents, and be compatible with printing processes. When combined with P-type organic semiconductors, high-performance complementary circuits can be formed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure and modeled charge carrier mobility of benzobis(thiadiazole) derivatives
Mamada M, et al.
New. J. Chem., 40(2), 1403-1411 (2016)
A Unique Solution-Processable n-Type Semiconductor Material Design for High-Performance Organic Field-Effect Transistors
Mamada M, et al.
Chemistry of Materials, 27(1), 141-147 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service