Skip to Content
Merck
All Photos(3)

Documents

767638

Sigma-Aldrich

Dinaphtho[2,3-b:2′,3′-f]thieno[3,2-b]thiophene

sublimed grade, 99%

Synonym(s):

DNTT, Naphtho[2,3-b]naphtho[2′,3′:4,5]thieno[2,3-d]thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12S2
CAS Number:
Molecular Weight:
340.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

99%

form

powder or crystals

mp

425-430 °C

semiconductor properties

P-type (mobility=2 cm2/V·s)

SMILES string

c1ccc2cc3c(cc2c1)sc4c5cc6ccccc6cc5sc34

InChI

1S/C22H12S2/c1-3-7-15-11-19-17(9-13(15)5-1)21-22(23-19)18-10-14-6-2-4-8-16(14)12-20(18)24-21/h1-12H

InChI key

CZWHMRTTWFJMBC-UHFFFAOYSA-N

General description

Dinaphtho[2,3-b:2′,3′-f]thieno[3,2-b]thiophene (DNTT) is a semiconducting polymer that has π-extended heteroarenes with six fused aromatic rings. It is a thermally stable crystal that has a hole mobility of 1 cm2V-1s-1 which can be used for a majority of electronic applications.

Application

It is mainly used as an organic semiconductor in the fabrication of organic field effect transistors (OFETs) for a variety of applications such as implantable electronics, large-area sensitive catheters, and light emitting diodes (LEDs).
Organic Field Effect Transistor (OFET) Materials; p-Type Organic Semiconductors; p-Type Small Molecules; sublimed grade materials

Legal Information

Product of Nippon Kayaku

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hongyan Wang et al.
PloS one, 8(11), e79118-e79118 (2013-11-19)
More and more transcription factors and their motifs have been reported and linked to specific gene expression levels. However, focusing only on transcription is not sufficient for mechanism research. Most genes, especially in eukaryotes, are alternatively spliced to different isoforms.
Martin Kaltenbrunner et al.
Nature, 499(7459), 458-463 (2013-07-28)
Electronic devices have advanced from their heavy, bulky origins to become smart, mobile appliances. Nevertheless, they remain rigid, which precludes their intimate integration into everyday life. Flexible, textile and stretchable electronics are emerging research areas and may yield mainstream technologies.
Contact Doping and Ultrathin Gate Dielectrics for Nanoscale Organic Thin-Film Transistors
Ante F, et al.
Small, 7(9), 1186-1191 (2011)
Mechanically adaptive organic transistors for implantable electronics
Reeder J, et al.
Advanced Materials, 26(29), 4967-4973 (2014)
Bishwajeet Singh Bhardwaj et al.
Materials (Basel, Switzerland), 12(4) (2019-02-20)
The application of dinaphthothienothiophene (DNTT) molecules, a novel organic semiconductor material, has recently increased due to its high charge carrier mobility and thermal stability. Since the structural properties of DNTT molecules, such as the molecular density distribution and molecular orientations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service