Skip to Content
Merck
All Photos(1)

Documents

90885

Sigma-Aldrich

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol

97%

Synonym(s):

1H,1H,2H,2H-Perfluoro-1-octanethiol, 1H,1H,2H,2H-Perfluorooctyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5CH2CH2SH
CAS Number:
Molecular Weight:
380.17
MDL number:
UNSPSC Code:
12352105
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (GC)
97%

form

liquid

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCS

InChI

1S/C8H5F13S/c9-3(10,1-2-22)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h22H,1-2H2

InChI key

GTPHVVCYEWPQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol is a fluorinated alkanethiol, which is also known as 1H,1H,2H,2H-Perfluorooctanethiol.

Application

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol is used to differentiate the effect on the work function and surface wetting for silver. Additionally, it can be used as a starting material to synthesize F-alkyl aroxysulfonyl carbamates and thiocarbamates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of F-alkyl N-sulfonyl carbamates and thiocarbamates
M Beji
Journal of Fluorine Chemistry, 99, 17-24 (1999)
Guangyu Rong et al.
Science advances, 6(33), eaaz1774-eaaz1774 (2020-08-28)
Cytosolic delivery of peptides remains a challenging task owing to their susceptibility to enzymatic degradation and the existence of multiple intracellular barriers. Here, we report a new strategy to address these issues by decoration of a fluorous tag on the
Concomitant tuning of metal work function and wetting property with mixed self-assembled monolayers
CY Chen
Organic Electronics, 12, 148-153 (2011)
Yong Jin Jeong et al.
ACS nano, 12(8), 7701-7709 (2018-07-20)
Photoresponsive transistor memories that can be erased using light-only bias are of significant interest owing to their convenient elimination of stored data for information delivery. Herein, we suggest a strategy to improve light-erasable organic transistor memories, which enables fast "photoinduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service