Skip to Content
Merck
All Photos(1)

Key Documents

532789

Sigma-Aldrich

1H,1H,2H,2H-Perfluoro-1-decanol

97%

Synonym(s):

3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanol, 1H,1H,2H,2H-Heptadecafluoro-n-decanol, 1H,1H,2H,2H-Perfluoro-1-decanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7CH2CH2OH
CAS Number:
Molecular Weight:
464.12
Beilstein:
2227487
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

functional group

fluoro
hydroxyl

SMILES string

OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10H5F17O/c11-3(12,1-2-28)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h28H,1-2H2

InChI key

JJUBFBTUBACDHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1H,1H,2H,2H-Perfluoro-1-decanol (8:2 FTOH) is an 8:2 fluorotelomer alcohol. The quantitative analysis of 8:2 FTOH in soil can be done using GC/MS with instrument detection limit (IDL) of 10fg/L. Studies indicate that hydroxyl radical causes indirect photodegradation of 8:2 FTOH in aqueous media.

Application

1H,1H,2H,2H-Perfluoro-1-decanol (8:2 FTOH) may be used in the preparation of 8:2 FTOH sulfate and 8:2 FTOH glucuronide.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgitte Lindeman et al.
Reproductive toxicology (Elmsford, N.Y.), 33(4), 531-537 (2011-05-03)
Testicular toxicity is observed following exposure of rats to per- and polyfluorinated compounds (PFCs). Such compounds were also shown to induce oxidative stress and changes in ABC efflux transporters e.g. P-gp, implying two mechanisms which may contribute to testicular toxicity.
Ning Wang et al.
Environmental science & technology, 39(2), 531-538 (2005-02-15)
This study investigated the biodegradation potential of 3-(14)C,1H,1H,2H,2H-perfluorodecanol [CF3(CF2)6(14)CF2CH2CH2OH, 14C-labeled 8-2 telomer B alcohol or 14C-labeled 8-2 TBA] by diluted activated sludge from a domestic wastewater treatment plant under aerobic conditions. After sample extraction with acetonitrile, biotransformation products were separated
Aqueous photolysis of 8: 2 fluorotelomer alcohol.
Gauthier SA and Mabury SA.
Environmental Toxicology and Chemistry / Setac, 24(8), 1837-1846 (2005)
Characterization of fluorinated metabolites by a gas chromatographic-helium microwave plasma detector--the biotransformation of 1H, 1H, 2H, 2H-perfluorodecanol to perfluorooctanoate.
D F Hagen et al.
Analytical biochemistry, 118(2), 336-343 (1981-12-01)
Suzanne A Gauthier et al.
Environmental toxicology and chemistry, 24(8), 1837-1846 (2005-09-13)
The 8:2 fluorotelomer alcohol (8:2 FTOH) was photodegraded in aqueous hydrogen peroxide solutions, synthetic field water (SFW) systems, and Lake Ontario (Canada) water samples. It was found to undergo indirect photolysis, with the data suggesting that the hydroxyl radical was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service