908169
Platinum on carbon
Evonik Noblyst® P2057 3% Pt
Synonym(s):
Pt/C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22
Recommended Products
form
powder
contains
~60% water
extent of labeling
3 wt. % loading (dry basis)
weight
Unit weight excludes weight of water
matrix
activated carbon support
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Evonik Noblyst® P2057 3% Pt is a platinum on carbon catalyst. It has been shown to be effective in the following reactions:
- hydrogenation of aromatic nitro groups
Other Notes
Unit weight is of net dry catalyst.
Legal Information
Manufactured by Evonik Degussa GmbH.
Noblyst is a registered trademark of Evonik Degussa GmbH
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
nwg
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service