Skip to Content
Merck
All Photos(2)

Documents

908029

Sigma-Aldrich

Palladium on carbon

Evonik Noblyst® P1141 10% Pd

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

contains

~50% water

extent of labeling

10 wt. % loading (dry basis)

weight

Unit weight excludes weight of water

matrix

activated carbon support

Looking for similar products? Visit Product Comparison Guide

Application

Evonik Noblyst® P1141 10% Pd is a palladium on carbon catalyst. It has been shown to be effective in the following reactions:
  • hydrogenation of alkenes and alkynes
  • hydrogenation of aromatic carbonyls to alkyls
  • hydrogenation of nitriles to amines and aldehydes
  • hydrogenation of aldehydes and ketones
  • hydrogenation of heteroaromatics
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Other Notes

Unit weight is of net dry catalyst.

Legal Information

Manufactured by Evonik Degussa GmbH.
Noblyst is a registered trademark of Evonik Degussa GmbH

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service