Skip to Content
Merck
All Photos(1)

Key Documents

905933

Sigma-Aldrich

Gold nanospheres

15 nm diameter, Cy3 and streptavidin functionalized, powder

Synonym(s):

Cy3 labeled gold nanospheres, Cy3 tagged gold nanospheres, Dye labeled gold nanoparticles, Fluorescent labeled gold nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanospheres
powder

diameter

15 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.

Legal Information

Product of Nanopartz Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescently Labeled Gold Nanoparticles with Minimal Fluorescence Quenching.
Lu Y, et al.
The Journal of Physical Chemistry C, 114, 17446-17446 (2010)
Byoungjin Kim et al.
Nature nanotechnology, 5(6), 465-472 (2010-04-13)
Nanoparticles have great potential as controllable drug delivery vehicles because of their size and modular functionality. Timing and location are important parameters when optimizing nanoparticles for delivery of chemotherapeutics. Here, we show that gold nanoparticles carrying either fluorescein or doxorubicin
Nuri Oh et al.
ACS nano, 8(6), 6232-6241 (2014-05-20)
Significant quantities of synthetic nanoparticles circulating in the body are cleared and retained for long periods of time in the resident macrophages of the mononuclear phagocytic system (MPS), increasing the likelihood of nanoparticle-mediated chronic toxicity. To date, there has been
Gold nanoparticles in chemical and biological sensing.
Krishnendu Saha et al.
Chemical reviews, 112(5), 2739-2779 (2012-02-03)
Xiaohua Huang et al.
Journal of the American Chemical Society, 128(6), 2115-2120 (2006-02-09)
Due to strong electric fields at the surface, the absorption and scattering of electromagnetic radiation by noble metal nanoparticles are strongly enhanced. These unique properties provide the potential of designing novel optically active reagents for simultaneous molecular imaging and photothermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service