Skip to Content
Merck

808237

Aluminum-doped zinc oxide nanoparticle ink

2.5 wt. %, viscosity 2.2 cP, work function -3.9 eV

Synonym(s):

AZO ink, Al-doped ZnO dispersion, Al-dopend ZnO suspension, Al:ZnO ink, Avantama N-21X, Nanograde N-21X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
12352103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

dispersion

concentration

2.5 wt. % (crystalline Al doped ZnO (3.15 mol% Al) in mixture of alcohols)

work function

-3.7--4.1 eV

particle size

8-16 nm

viscosity

1.7-2.7 cP

Application

Al-doped ZnO nanoparticle ink is for spin coating and doctor blading for the use as electron transport layer in printed electronics. Al-doped ZnO nanoparticle ink is universally applicable in normal and inverted architecture with performance comparable to evaporated calcium.

Preparation Note

  • Storage: In dark at room temperature.
  • Prior to application: Shake, ultrasonicate with sonic horn and (optionally) filter through 0.45μm PTFE filter.
  • Post-treatment: Annealing of deposited Al doped ZnO films at 80°C - 120°C.

Legal Information

Product of Avantama Ltd.


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

60.8 °F

flash_point_c

16 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library