Skip to Content
Merck
All Photos(1)

Key Documents

902942

Sigma-Aldrich

Graphite dispersion

few-layers (3-15 graphene layers), 1.0mg/mL in acetone-water, avg. no. of layers, 3 ‑ 15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141908
NACRES:
NA.23

description

Capacity:400−450 mAh/g
Conductivity: 1.0x10-3 − 1.0x10-5 S/m
I_D/I_G peaks ratio: <0.25 (by Raman)
Surface area: 1.0x105–4.0x106 mm²/g

form

dispersion

feature

avg. no. of layers 3 ‑ 15

composition

C: >96%, H: 0 −1%, N: 0 −1%, Oxygen: 0−2% (graphene)

General description

Graphite dispersion is an acceptor molecule with high thermal and electrical conductivity. It can be used in the formation of an electrode for electrochemical applications.

Application

Graphite dispersion (few-layers: 3-15 graphene layers, 1.0mg/mL in acetone-water) can be used for a variety application such as
  • in the synthesis of graphene layer
  • in the preparation of ruthenium catalyst for the synthesis of ammonia
  • development of biosensors
  • fabrication of multi-layer thin films for li-ion micro-batteries

Preparation Note

Production method: Thermomechanical exfoliation of natural graphite in a liquid medium controlled at a nanoscale level.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Autonomous microscopic bunch inspection using region-based deep learning for evaluating graphite powder dispersion
Liu H, et al.
Construction Building Materials, 173, 525-539 (2018)
Preparation of efficient ruthenium catalysts for ammonia synthesis via high surface area graphite dispersion
Han W, et al.
Reaction Kinetics, Mechanisms and Catalysis, 113(2), 361-374 (2014)
Development of flexible sensors using knit fabrics with conductive polyaniline coating and graphite electrodes
de Oliveira CRS, et al.
Journal of Applied Polymer Science, 134(18), 525-539 (2017)
Preparation and characterization of graphite composites of polyaniline
Ozerol EA, et al.
Microelectronic Engineering, 146, 76-80 (2015)
Synthesis of graphene layers using graphite dispersion in aqueous surfactant solutions
Sim Y, et al.
The Journal of the Korean Physical Society, 58(4), 938-942 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service