Skip to Content
Merck
All Photos(3)

Key Documents

902500

Sigma-Aldrich

SHT-263 Solarpur®

greener alternative

Synonym(s):

N2,N2,N2′,N2′,N7,N7,N7′,N7′-octakis(4-methoxyphenyl)-9,9′-spirobi[9H-fluorene]-2,2′,7,7′-tetramine, Spiro-MeOTAD, Spiro-OMeTAD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H68N4O8
CAS Number:
Molecular Weight:
1225.43
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥99.9%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.1% Residual solvent

mp

247 °C

greener alternative category

SMILES string

N(c%14ccc(cc%14)OC)(c%13ccc(cc%13)OC)c1cc2c(cc1)c3c(cc(cc3)N(c%12ccc(cc%12)OC)c%11ccc(cc%11)OC)C42c5c(ccc(c5)N(c%10ccc(cc%10)OC)c9ccc(cc9)OC)c6c4cc(cc6)N(c8ccc(cc8)OC)c7ccc(cc7)OC

InChI

1S/C81H68N4O8/c1-86-65-29-9-53(10-30-65)82(54-11-31-66(87-2)32-12-54)61-25-45-73-74-46-26-62(83(55-13-33-67(88-3)34-14-55)56-15-35-68(89-4)36-16-56)50-78(74)81(77(73)49-61)79-51-63(84(57-17-37-69(90-5)38-18-57)58-19-39-70(91-6)40-20-58)27-47-75(79)76-48-28-64(52-80(76)81)85(59-21-41-71(92-7)42-22-59)60-23-43-72(93-8)44-24-60/h9-52H,1-8H3

InChI key

XDXWNHPWWKGTKO-UHFFFAOYSA-N

General description

Both SHT-263S and SHT-263 can be offered in bulk quantities.
SHT-263 Solarpur® is an organic spiro molecule that is used as a hole transporting material (HTM). Its properties include high stability, good solubility, and an amorphous structure. It is majorly used in the fabrication of high-performance solar cells.
We are committed to bringing you Greener Alternative Products,which adhere to one or more of The 12 Principles of Greener Chemistry. This product is an enabling product used as a Hole Transport Material for high-performance solar cells and thus has been enhanced for energy efficiency. Click here for more information.

Application

SHT-263 Solarpur® is a spiro based hole transporting material (HTM) with a HOMO level of -5.2 eV and a LUMO level of -2.3 eV. It is mainly used in the fabrication of perovskite-based solar cell.

Legal Information

Solarpur is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Metal- Free Porphyrins as Hole- Transporting Materials in Mesoporous Perovskite Solar Cells.
Sygkridou D, et al.
ChemistrySelect, 3(9), 2536-2541 (2018)
Unique hole transport layer of PCDTBT doped with PCPDTBT for perovskite solar cells
Pratyusha T, et al.
Materials Today: Proceedings, 4(7), 6820-6826 (2017)
Air-exposure induced dopant redistribution and energy level shifts in spin-coated spiro-MeOTAD films
Hawash Z, et al.
Chemistry of Materials, 27(2), 562-569 (2015)
Unique Hole Transport Layer of PCDTBT Doped with PCPDTBT for Perovskite Solar Cells
Pratyusha T, et al.
Materials Today, 4(7), 6820-6826 (2017)
Hairen Tan et al.
Science (New York, N.Y.), 355(6326), 722-726 (2017-02-06)
Planar perovskite solar cells (PSCs) made entirely via solution processing at low temperatures (<150°C) offer promise for simple manufacturing, compatibility with flexible substrates, and perovskite-based tandem devices. However, these PSCs require an electron-selective layer that performs well with similar processing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service