Skip to Content
Merck
All Photos(1)

Key Documents

901591

Sigma-Aldrich

18-Crown-6 solution

1.0 M in THF

Synonym(s):

1,4,7,10,13,16-Hexaoxacyclooctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H24O6
CAS Number:
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF

density

0.944 g/mL

SMILES string

O1CCOCCOCCOCCOCCOCC1

InChI

1S/C12H24O6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2

InChI key

XEZNGIUYQVAUSS-UHFFFAOYSA-N

Application

18-Crown-6 may be used to catalyze the N-alkylation of heterocyclic compounds and allylation of functionalized aldehydes.
Additive for greener etherification using KF-alumina.
Phase-transfer catalyst used in a chemoselective reduction of fused tetrazoles with NaBH4 and potassium hydroxide.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.0 °F

Flash Point(C)

-18.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 3275-3275 (2006)
Phase-transfer alkylation of heterocycles in the presence of 18-crown-6 and potassium tert-butoxide.
Guida WC and Mathre DJ
The Journal of Organic Chemistry, 45(16), 3172-3176 (1980)
Preparation and purification of 18-crown-6 [1, 4, 7, 10, 13, 16-hexaoxacyclooctadecane].
Gokel GW, et al.
The Journal of Organic Chemistry, 39(16), 2445-2446 (1974)
Hiroaki Kotani et al.
Journal of the American Chemical Society, 133(29), 11092-11095 (2011-06-28)
Addition of potassium superoxide with 18-crown-6 ether (KO(2)(•-)-18-crown-6) to a toluene solution of an acridinium ion-linked porphyrin triad (Acr(+)-H(2)P-Acr(+)) resulted in a remarkable enhancement of the fluorescence intensity. Thus, Acr(+)-H(2)P-Acr(+) acts as an efficient fluorescence sensor for superoxide. Electron transfer
Shih-Wey Yeh et al.
Inorganic chemistry, 51(7), 4076-4087 (2012-03-13)
The reversible redox transformations [(NO)(2)Fe(S(t)Bu)(2)](-) ⇌ [Fe(μ-S(t)Bu)(NO)(2)](2)(2-) ⇌ [Fe(μ-S(t)Bu)(NO)(2)](2)(-) ⇌ [Fe(μ-S(t)Bu)(NO)(2)](2) and [cation][(NO)(2)Fe(SEt)(2)] ⇌ [cation](2)[(NO)(2)Fe(SEt)(2)] (cation = K(+)-18-crown-6 ether) are demonstrated. The countercation of the {Fe(NO)(2)}(9) dinitrosyliron complexes (DNICs) functions to control the formation of the {Fe(NO)(2)}(10){Fe(NO)(2)}(10) dianionic reduced Roussin's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service