Skip to Content
Merck
All Photos(3)

Documents

158399

Sigma-Aldrich

Dibenzo-18-crown-6

98%

Synonym(s):

2,3,11,12-Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene, 6,7,9,10,17,18,20,21-Octahydrodibenzo[b,k][1,4,7,10,13,16]hexaoxacyclooctadecene, DB18C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O6
CAS Number:
Molecular Weight:
360.40
Beilstein:
1162153
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

162-164 °C (lit.)

SMILES string

C1COc2ccccc2OCCOCCOc3ccccc3OCCO1

InChI

1S/C20H24O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h1-8H,9-16H2

InChI key

YSSSPARMOAYJTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzo-18-crown-6 is a macrocyclic ligand used in the selective binding and extraction of alkali metal cations in solution phase.

Application

Phase-transfer catalyst employed in monoazaporphyrin syntheses. Agent used for ion transfer across membranes and as a synthon for preparation of liquid crystal polyesters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding selectivity of dibenzo-18-crown-6 for alkali metal cations in aqueous solution: A density functional theory study using a continuum solvation model
Chang Min C et al.
Chemistry Central Journal, 6, 1-8 (2012)
Tetrahedron Letters, 36, 1567-1567 (1995)
Solvent influence upon complex formation between dibenzo-18-crown-6 with the Y3+ metal cation in pure and binary mixed organic solvents
GH Rounaghi et al.
Journal of Chemical and Engineering Data, 56, 2836-2840 (2011)
Acta Polym., 45, 308-308 (1994)
Chang Min Choi et al.
The journal of physical chemistry. A, 114(42), 11167-11174 (2010-10-22)
Ultraviolet photodepletion spectra of dibenzo-18-crown-6-ether complexes with alkaline earth metal divalent cations (A(2+)-DB18C6, A = Ba, Sr, Ca, and Mg) were obtained in the gas phase using electrospray ionization quadrupole ion-trap reflectron time-of-flight mass spectrometry. Each spectrum exhibits the lowest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service