Skip to Content
Merck
All Photos(2)

Key Documents

901243

Sigma-Aldrich

4-(Acetylamino)phenyl]imidodisulfuryl difluoride

≥98%

Synonym(s):

(4-Acetamidophenyl)(fluorosulfonyl)sulfamoyl fluoride, AISF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8F2N2O5S2
CAS Number:
Molecular Weight:
314.29
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder or crystals

mp

146-149 °C

functional group

amide

SMILES string

F[S](=O)(=O)N([S](=O)(=O)F)c1ccc(cc1)NC(=O)C

InChI key

SMMIWAIOWMUFCR-UHFFFAOYSA-N

Application

4-(Acetylamino)phenyl]imidodisulfuryl difluoride (AISF) is a shelf-stable, crystalline reagent for synthesis of fluorosulfates and sulfamoyl fluorides via reaction with phenol and amine nucleophiles. A convenient alternative to the toxic sulfuryl fluoride gas traditionally required for installation of the valuable -SO2F functional group, the solid reagent AISF will find wide utility in preparation of synthetic precursors as well as chemical biology, polymer chemistry, cross-coupling, and deoxyfluorination applications.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Zhou et al.
Organic letters, 20(3), 812-815 (2018-01-13)
The design, synthesis, and application of [4-(acetylamino)phenyl]imidodisulfuryl difluoride (AISF), a shelf-stable, crystalline reagent for the synthesis of sulfur(VI) fluorides, is described. The utility of AISF is demonstrated in the synthesis of a diverse array of aryl fluorosulfates and sulfamoyl fluorides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service