900679
DOTA-functionalized silica gel
≥99%
Synonym(s):
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid-functionalized silica gel, Si-DOTA, SiliaMetS® DOTA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161600
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
form
powder
shelf life
2 yr (expiration date period)
reaction suitability
reagent type: catalyst
reagent type: chelator
surface area
480-550 m2/g
pore size
60 Å pore diameter
bulk density
679 g/L (typical tap density)
Application
Metal scavenger: Ca, Ce, Cu, Fe, Gd, La, Ni, Pd, Zn.
Caution
For prolonged storage, recommended conditions are to store product in a cool place (<8 °C) under inert atmosphere (argon) when possible.
Compatibility
This product is compatible with all common aqueous and organic solvents.
Legal Information
SiliaMetS is a registered trademark of Silicycle, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service