Skip to Content
Merck
All Photos(1)

Key Documents

900459

Sigma-Aldrich

Gold nanoparticles

10 nm, maleimide functionalized, conjugation kit

Synonym(s):

Au nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141717
NACRES:
NA.23

form

nanoparticles

packaging

pkg of 1 ea

particle size

10 nm

λmax

515-520 nm

functional group

maleimide

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanoparticles, 10 nm, maleimide functionalized, conjugation kit is a comprehensive kit that enables easy, one-step conjugation of your thiol-containing ligands to 10 nm gold nanoparticles. The kit includes pre-made mixtures of lyophilized (freeze-dried) maleimide gold nanoparticles, along with all the necessary materials for three independent conjugation reactions. The kit components comprise:
•3 vials of 10 nm Malemide-Activated Gold Nanoparticles (lyophilized)
•Protein Resuspension Buffer - 1.5ml
•Reaction Buffer - 1.5ml
•Quencher Solution - 1.5ml
•Reaction protocol
The gold nanoparticles in this kit are supplied as a lyophilized (freeze-dried) mixture. The conjugation reaction is initiated by simply reconstituting the freeze-dried nanoparticles with the protein of interest, which attaches to the nanoparticle surface via cysteine residues.
The gold conjugation procedure has a quick hands-on time of around 10 minutes, and the conjugate is fully ready for use within 3 hr. Researchers only need to pipette the biomolecule into a vial containing the nanoparticles, quench the reaction, and perform product washing through microcentrifugation.
The provided protocol is optimized for highly efficient covalent conjugation, resulting in stable gold nanoparticle conjugates. Optional protocol steps involve additional materials, such as 10% aqueous BSA (126615) and Tris saline resuspension buffer (PPB002), which are sold separately.

Application

Biocompatibility, facile conjugation to biomolecules and unique optical properties arising from surface plasmon resonance render the nanoparticles of gold useful for variety of biomedical research such as bio-imaging, bio-sensing, photothermal hyperthermia, drug delivery etc. Our maleimide functionalized gold nanoparticles kits contain ready-to-use mixtures and have been optimized for high efficiency one-step conjugation of thiol-modified ligands such as oligonucleotides, antibodies, antibody fragments, proteins and peptides.
This gold conjugation kit streamlines the process of conjugating your proteins or biomolecules to 10 nm gold nanoparticles. Gold conjugation helps researchers to visualize and assay a specific protein or antibody of interest. Gold conjugates are useful in a wide range of applications like lateral flow testing, immunoblotting, immuno-electron microscopy, immunohistochemical staining, and other bio-assays.
Different sizes of gold nanoparticles perform better in specific applications. For lateral flow and flow cytometry, we recommend using gold nanoparticles with diameters between 20 nm and 100 nm. For immunoblotting or immuno-electron microscopy, the optimal range is 5 nm to 40 nm. For bioassays utilizing dynamic light scattering, gold nanoparticles with diameters ranging from 60 nm to 100 nm are suggested.

Features and Benefits

•The resulting covalent conjugates offer greater stability compared to those prepared using passive adsorption methods.
•Fast and convenient conjugation reaction with no need for pre-activation.
•A spacer between the gold nanoparticle surface and the conjugated ligand minimizes the impact on the tertiary protein structure. This reduces the likelihood of poor performing conjugates, a common issue encountered with conjugates prepared by passive adsorption.
•Gold surface coating is precisely engineered to minimize non-specific protein binding in biological assays.

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service