Skip to Content
Merck
All Photos(2)

Key Documents

900278

Sigma-Aldrich

AdCyBrettPhos

Synonym(s):

2-(Adamantylcyclohexylphosphino)-2’,4’,6’-triisopropyl-3,6-dimethoxybiphenyl, Cyclohexyl adamantyl(2’,4’,6’-triisopropyl-3,6-dimethoxybiphenyl-2-yl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H57O2P
CAS Number:
Molecular Weight:
588.84
UNSPSC Code:
12352128
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

221-224 °C

functional group

phosphine

SMILES string

O(C1=CC=C(OC)C(=C1C=2C(=CC(=CC2C(C)C)C(C)C)C(C)C)P(C3CCCCC3)C45CC6CC(CC(C6)C4)C5)C

InChI

InChI=1S/C39H57O2P/c1-24(2)30-19-32(25(3)4)36(33(20-30)26(5)6)37-34(40-7)14-15-35(41-8)38(37)42(31-12-10-9-11-13-31)39-21-27-16-28(22-39)18-29(17-27)23-39/h14-15,19-20,24-29,31H,9-13,16-18,21-23H2,1-8H3

InChI key

AMESLWUOCNKBCL-UHFFFAOYSA-N

Application

AdCyBrettPhos can be used as a ligand in the Pd-catalyzed cross-coupling reactions.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan P King et al.
Organic letters, 23(20), 7927-7932 (2021-10-07)
The utilization of isolated Palladium Oxidative Addition Complexes (OACs) has had a significant impact on Pd-catalyzed and Pd-mediated cross-coupling reactions. Despite their importance, widespread utility of OACs has been limited by the instability of their precursor complexes. Herein, we report
Hong Zhang et al.
Organic letters, 20(6), 1580-1583 (2018-02-24)
Two catalyst systems are described, which together provide mild and general conditions for the Pd-catalyzed C-O cross-coupling of primary alcohols. For activated substrates, such as electron-deficient aryl halides, the commercially available ligand L2 promotes efficient coupling for a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service