Skip to Content
Merck
All Photos(1)

Documents

88424

Sigma-Aldrich

3-(2-Thienyl)-L-alanine

≥98.0% (TLC)

Synonym(s):

(S)-α-Amino-2-thiophenepropionic acid, (S)-2-Amino-3-(2-thienyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
Beilstein:
82872
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]20/D −30.5±1.5°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

255-263 °C (dec.) (lit.)
~260 °C (dec.)

application(s)

peptide synthesis

SMILES string

N[C@@H](Cc1cccs1)C(O)=O

InChI

1S/C7H9NO2S/c8-6(7(9)10)4-5-2-1-3-11-5/h1-3,6H,4,8H2,(H,9,10)/t6-/m0/s1

InChI key

WTOFYLAWDLQMBZ-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D P Bedard et al.
Journal of bacteriology, 141(1), 100-105 (1980-01-01)
When treated with the amino acid analog beta-2-DL-thienylalanine, cells of the yeast Saccharomyces cerevisiae accumulated in the G1 portion of the cell cycle at the "start" event. This G1 arrest was accompanied by a rapid decrease in the rate of
"Physiological phenylketonuria": a biochemical defect caused by delayed maturation of the phenylalanine hydroxylation pathway and by competition with the phenylalanine biosynthetic pathway.
K J Brown
Medical hypotheses, 6(2), 215-224 (1980-02-01)
Uptake and incorporation of amino acids by suspension cultured mammalian cells: a comparative study involving eleven naturally-occurring and four analogue amino acids.
D N Wheatley et al.
Cytobios, 30(118), 101-126 (1981-01-01)
G Riccardi et al.
Journal of bacteriology, 147(3), 1002-1007 (1981-09-01)
Mutants of Spirulina platensis resistant to 5-fluorotryptophan, beta-3-thienyl-alanine, ethionine, p-fluorophenylalanine, or azetidine-2-carboxylic acid were isolated. Some of these mutants appeared to be resistant to more than one analog and to overproduce the corresponding amino acids. A second group was composed
K J Brown et al.
Microbiology and immunology, 24(7), 603-615 (1980-01-01)
Growth comparisons were made, using Shigella, Escherichia, and Salmonella cultures, in liquid and agar-solidified defined media containing beta-2-thienylalanine (beta-2-t). The comparisons were performed to determine the nature of growth inhibition by beta-2-t under different physical growth conditions. In a plate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service