
3-(2-Thienyl)-L-alanine

Synonym(s):
(S)-α-Amino-2-thiophenepropionic acid, (S)-2-Amino-3-(2-thienyl)propionic acid
Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing