Skip to Content
Merck
All Photos(1)

Key Documents

87306

Sigma-Aldrich

Tetrahexylammonium hydroxide solution

~40% in H2O (T)

Synonym(s):

Tetra(n-hexyl)ammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]4N(OH)
CAS Number:
Molecular Weight:
371.68
Beilstein:
6019685
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

solution

Quality Level

concentration

~40% in H2O (T)

impurities

≤0.1% halides (as bromide)

anion traces

sulfate (SO42-): ≤1000 mg/kg

functional group

amine

SMILES string

[OH-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C24H52N.H2O/c1-5-9-13-17-21-25(22-18-14-10-6-2,23-19-15-11-7-3)24-20-16-12-8-4;/h5-24H2,1-4H3;1H2/q+1;/p-1

InChI key

JCJNUSDBRRKQPC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetrahexylammonium hydroxide is a phase-transfer reagent that can be used:
  • To synthesize β-D-xyloside- and β-D-xylobioside-based ionic liquids containing tetrahexylammonium cation.
  • As an alkali-free gelling agent to prepare amorphous microporous silica-alumina.
  • To synthesize tetrahexylammonium prolinate, a promising candidate to prepare a solid supported ionic liquid phase (SILP) carbon dioxide absorber.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of diuretic agents in doping control.
Ventura R and Segura J
Journal of Chromatography. B, Biomedical Sciences and Applications, 687(1), 127-144 (1996)
Mesoporous silica-aluminas as catalysts for the alkylation of aromatic hydrocarbons with olefins.
Perego C, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 27(2-3), 345-354 (1999)
CO 2 Capture technologies: Current status and new directions using supported ionic liquid phase (SILP) absorbers.
Kolding H, et al.
Science China: Chemistry, 55(8), 1648-1656 (2012)
Preparation of new ?-D-xyloside-and ?-D-xylobioside-based ionic liquids through chemical and/or enzymatic reactions.
Gatard S, et al.
Carbohydrate Research, 451, 72-80 (2017)
The effects of tetrahexylammonium chloride on calcium mobilization from cerebellar microsomes.
L G Sayers et al.
Biochemical Society transactions, 21(2), 105S-105S (1993-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service