Skip to Content
Merck
All Photos(1)

Key Documents

85630

Sigma-Aldrich

Squalane

technical, ≥90% (GC)

Synonym(s):

2,6,10,15,19,23-Hexamethyltetracosane, Cosbiol, Perhydrosqualene, Robane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)CH2CH2-]2
CAS Number:
Molecular Weight:
422.81
Beilstein:
776019
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.452 (lit.)
n20/D 1.452

bp

176 °C/0.05 mmHg (lit.)

mp

−38 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

InChI

1S/C30H62/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h25-30H,9-24H2,1-8H3

InChI key

PRAKJMSDJKAYCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Squalane, a saturated hydrocarbon, serves as an aprotic and inert solvent in organic synthesis. Its fully saturated structure makes it ideal for radical reactions. Additionally, squalane is used as a stationary phase in gas chromatography (GC)

Application

  • Thixotropic Molecular Oleogels: Squalane is utilized in the development of thixotropic molecular oleogels with alkylanilide gelators, enhancing the material′s stability and usability in various pharmaceutical and cosmetic applications (Ohsedo, 2023).
  • Mucoadhesive Pickering Nanoemulsions: This compound supports the creation of mucoadhesive Pickering nanoemulsions that leverage dynamic covalent chemistry, offering advanced delivery mechanisms for active ingredients in pharmaceutical formulations (Smallridge et al., 2023).
  • Enhancement of Rheological Properties: Squalane is applied in studying the rheological properties of small-molecular liquids at high shear strain rates, which is crucial for improving the processing and application of consumer products (Jadhao, 2023).
  • Stabilization of Emulsion Systems: It is also used in emulsion systems as part of a sucrose fatty acid ester-mixed emulsifier, enhancing the stability and performance of food products and cosmetics (Hu et al., 2023).
  • Thermophysical Properties of Alkanes: Research includes the comparison of force fields for predicting the thermophysical properties of long linear and branched alkanes, with squalane serving as a key example in these studies, important for chemical engineering and material science (Stephan et al., 2023).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

424.4 °F - closed cup

Flash Point(C)

218 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-phase methanol synthesis in apolar (squalane) and polar (tetraethylene glycol dimethylether) solvents
van der Laan et al.
Catalysis Today, 48, 93-100 (1999)
A gas chromatography-mass spectrometry method for quantifying squalane and squalene in aerosol emissions of electronic cigarette, or vaping, products
Cowan et al.
Talanta, 238, 122985-122985 (2022)
Hydroxyl radical-initiated aging of particulate squalane
Zhang et al.
Atmospheric Environment, 237, 117663-117663 (2020)
Shinya Kato et al.
Journal of biomedical nanotechnology, 6(1), 52-58 (2010-05-27)
We dissolved fullerene-C60 in squalane (LipoFullerene; LF-SQ, C60-eq.: 500 ppm) and examined its defensive effects against 2,4-nonadienal (NDA)-induced cell injury in HaCaT keratinocytes and wrinkle formation in three dimensional (3D)-human skin tissue model. NDA is an analog of 4-hydroxynonenal, one
Scott Yockel et al.
Topics in current chemistry, 307, 43-67 (2011-04-21)
In this chapter we describe molecular dynamics simulation methods in which the system being studied is divided into a region where quantum mechanics (QM) is used to determine forces for doing Born-Oppenheimer direct dynamics calculations (i.e., doing electronic structure calculations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service