Skip to Content
Merck
All Photos(1)

Key Documents

850993

Sigma-Aldrich

2-Aminoisobutyric acid

98%, for peptide synthesis

Synonym(s):

α-Aminoisobutyric acid, 2-Methylalanine, Aib

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
506496
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product name

2-Aminoisobutyric acid, 98%

Assay

98%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

≥300 °C

application(s)

peptide synthesis

SMILES string

CC(C)(N)C(O)=O

InChI

1S/C4H9NO2/c1-4(2,5)3(6)7/h5H2,1-2H3,(H,6,7)

InChI key

FUOOLUPWFVMBKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Aminoisobutyric acid also known as α-aminobutyric acid, is an amino acid used in solution-phase peptide synthesis. It is a desirable building block for peptides because of its strong tendency to cause the peptide to form a helical shape.

Application

2-Aminoisobutyric acid can be used to synthesize self-assembled polypeptide nanoparticles. Incorporation of this compound into the peptide chain can prevent undesired reactions since it is di-α-substituted, and inert to C−H abstraction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical methods for peptide macrocyclisation
Chemistry?A European Journal , 27, 69-88 (2021)
Amphiphilic polypeptides with prolonged enzymatic stability for the preparation of self-assembled nanobiomaterials
Royal Society of Chemistry Advances, 8, 34603-34613 (2018)
C Lucarelli et al.
Journal of chromatography, 541(1-2), 285-296 (1991-03-22)
A procedure is described for the determination of alpha-methyldopa (MD) [L-3-(3,4-dihydroxyphenyl)-2-methylalanine], its metabolite and catecholamines in the urine and plasma of patients undergoing MD therapy, by high-performance liquid chromatography with dual working electrode coulometric detection. An efficient sample preparation procedure
Jamie E Elsila et al.
Astrobiology, 11(2), 123-133 (2011-03-23)
The presence of nonprotein α-dialkyl-amino acids such as α-aminoisobutyric acid (α-AIB) and isovaline (Iva), which are considered to be relatively rare in the terrestrial biosphere, has long been used as an indication of the indigeneity of meteoritic amino acids. However
Evgeniy Salnikov et al.
Journal of biomolecular NMR, 45(4), 373-387 (2009-10-14)
In protein NMR spectroscopy the chemical shift provides important information for the assignment of residues and a first structural evaluation of dihedral angles. Furthermore, angular restraints are obtained from oriented samples by solution and solid-state NMR spectroscopic approaches. Whereas the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service