Skip to Content
Merck
All Photos(1)

Key Documents

81350

Sigma-Aldrich

Polypropylene glycol

P 400

Synonym(s):

1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

liquid

mol wt

Mn 446

refractive index

n20/D 1.447

viscosity

~100 mPa.s(20 °C)

solubility

water: miscible (completely)

density

1.01 g/mL at 20 °C

SMILES string

O(C(CO)C)CC(O)C

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylene glycol is an aliphatic alcohol. It is an addition polymer of propylene glycol and water represented as H[OCH3]nOH in which n represents the average number of oxypropylene groups.

Application

Propylene glycol (PPG) may be used as a viscosity decreasing agent, a solvent and a fragrance ingredient in cosmetics. PPG may be used as a good swelling agent for the synthesis of large pore mesoporous materials.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterization of organo-modified SBA-15 by using polypropylene glycol as a swelling agent
Park BG, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 66(2), 229-238 (2003)
Anup Aggarwal et al.
Cell, 170(2), 249-259 (2017-07-04)
Widespread resistance to first-line TB drugs is a major problem that will likely only be resolved through the development of new drugs with novel mechanisms of action. We have used structure-guided methods to develop a lead molecule that targets the
L Hong et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 83(6 Pt 1), 061508-061508 (2011-07-30)
There are various arguments and models connecting the characteristic length associated with the boson peak vibrations ξ to the length scale of dynamical heterogeneity L(het). ξ is usually defined as the ratio of the transverse sound velocity to the boson
Izzuddin Zaman et al.
Nanoscale, 4(15), 4578-4586 (2012-06-19)
In spite of extensive studies conducted on carbon nanotubes and silicate layers for their polymer-based nanocomposites, the rise of graphene now provides a more promising candidate due to its exceptionally high mechanical performance and electrical and thermal conductivities. The present
R De Lisi et al.
Physical chemistry chemical physics : PCCP, 13(27), 12571-12577 (2011-06-15)
The study highlighted the main forces driving the formation of hydroxypropyl-cyclodextrins (HP-CDs) + poly(propylene) glycol 725 g mol(-1) inclusion complexes. The temperature parameter was chosen as the variable to modulate the hydrophobicity of the polymer, and consequently ITC experiments as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service