Skip to Content
Products
Cart
0
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Polypropylene glycol
Synonym(s):
PPG, Poly(propylene oxide)
Linear Formula:
H[OCH(CH
3
)CH
2
]
n
OH
CAS Number:
25322-69-4
Products
Brand
Sigma-Aldrich (8)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
81380
Polypropylene glycol,
P 2,000
Expand
View Pricing
81350
Polypropylene glycol,
P 400
Expand
View Pricing
202304
Polypropylene glycol,
average M
n
~425
Expand
View Pricing
202320
Polypropylene glycol,
average M
n
~1,000
Expand
View Pricing
202339
Polypropylene glycol,
average M
n
~2,000
Expand
View Pricing
202355
Polypropylene glycol,
average M
n
~4,000
Expand
View Pricing
202312
Polypropylene glycol,
average M
n
~725
Expand
View Pricing
202347
Polypropylene glycol,
average M
n
~2,700
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Français - FR
Learn More