Skip to Content
Merck
All Photos(2)

Key Documents

806447

Sigma-Aldrich

2,3,4,5,6-Pentafluoroanilino(oxo)acetic acid

Synonym(s):

[(2,3,4,5,6-Pentafluorophenyl)amino](oxo)acetic acid, [(2,3,4,5,6-Pentafluorophenyl)carbamoyl]formic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2F5NO3
CAS Number:
Molecular Weight:
255.10
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

Quality Level

form

powder

SMILES string

O=C(C(O)=O)NC1=C(F)C(F)=C(F)C(F)=C1F

InChI

1S/C8H2F5NO3/c9-1-2(10)4(12)6(5(13)3(1)11)14-7(15)8(16)17/h(H,14,15)(H,16,17)

InChI key

JHFLRXFZPBDINI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service