Skip to Content
Merck
All Photos(1)

Key Documents

805491

Sigma-Aldrich

2-Fluoro-6-(piperidine-1-sulfonyl)anilino(oxo)acetic acid

Synonym(s):

[(2-Fluoro-6-(1-piperidinylsulfonyl)phenyl)carbamoyl]formic acid, [(2-Fluoro-6-(piperidine-1-sulfonyl)phenyl)amino](oxo)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15FN2O5S
Molecular Weight:
330.33
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

95% (HPLC)

Quality Level

form

powder

SMILES string

O=S(N1CCCCC1)(C2=CC=CC(F)=C2NC(C(O)=O)=O)=O

InChI

1S/C13H15FN2O5S/c14-9-5-4-6-10(11(9)15-12(17)13(18)19)22(20,21)16-7-2-1-3-8-16/h4-6H,1-3,7-8H2,(H,15,17)(H,18,19)

InChI key

WFXNBMKFCASFSD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service