Skip to Content
Merck
All Photos(1)

Key Documents

805173

Sigma-Aldrich

Green Fluorescent PLGA microspheres

25 μm average diameter

Synonym(s):

Green Fluorescent PLGA nanoparticles, Degradex® Fluorescent PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)x(C2H2O2)y
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

description

Excitation/emission: 460/500 nm

Quality Level

form

powder

average diameter

25 μm

density

1.3 g/cm3

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Degradex® is made with poly(D,L-lactide-co-glycolide) with an L/G ratio of 50/50 and a Mw of 30,000.
To re-constitute:
  • Remove the product from freezer
  • Wait until temperature rises to the ambient temperature
  • Add DI water or buffer to the container
  • Vortex and sonicate for 5 minutes to ensure the microspheres are well suspended.

Application

Fluorescent PLGA particles for imaging and cell tracking in drug discovery. Particles can be used for fluorscent assays and drug delivery.

Legal Information

Degradex is a registered trademark of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service