Skip to Content
Merck
All Photos(1)

Key Documents

L4655

Sigma-Aldrich

Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green

aqueous suspension, 1.0 μm mean particle size

Synonym(s):

Fluorescent Latex Beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

form

aqueous suspension

composition

Solids, 2.5%

technique(s)

cell based assay: suitable

mean particle size

1.0 μm

fluorescence

λex ~470 nm; λem ~505 nm

suitability

suitable for immunoassay

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

Application

Carboxylate-modified polystyrene latex beads have been used to collect morphofunctional data about the immune systems of Carabus lefebvrei, a helicophagous Italian endemic ground beetle often used as an indicator of the habitat quality of the Apennines mountain forests. Carboxylate-modified polystyrene latex beads have also been used to inform comparative studies on immunity defense mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa and to investigate the phagocytic activity of cultured rat thyroid cells.
Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green has been used:
  • in epidermal growth factor conjugate preparation for indirect immunofluorescence imaging studies
  • for culturing mouse bronchoalveolar macrophages and human monocyte-derived macrophages prior to phagocytosis imaging studies
  • in coadministration with Salmonella expressed green fluorescent protein (GFP) for confocal microscopy studies and in biofluorescence imaging

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Libin Huang et al.
Ocular immunology and inflammation, 24(6), 684-692 (2016-12-10)
To evaluate the effect of CX3CL1/CX3CR1 signaling on the interaction between mesenchymal stem cells (MSCs) and retinal microglia. Supernatants of homogenized retina were harvested from light-damaged SD rats (ISHR) to stimulated retinal microglia. Stimulated microglia were cocultured with MSCs, CX3CL1
Elin Silverpil et al.
Inflammation, 34(1), 1-9 (2010-03-27)
There is now substantial evidence that the cytokine interleukin-17 orchestrates the accumulation of neutrophils in mammals and thereby contributes to host defense. However, the role of IL-17 in controlling neutrophil turnover is not fully understood. Here, we demonstrate that IL-17
Po-Tsang Lee et al.
Animals : an open access journal from MDPI, 11(8) (2021-08-28)
We investigated whether phycoerythrin (PE), a pigment sourced from marine algae, could act as an immunomodulatory agent in whiteleg shrimp (Litopenaeus vannamei). To this end, PE was extracted and purified from a PE-rich macroalgae, Colaconema sp. Our in vitro analysis
J Ablin et al.
Apoptosis : an international journal on programmed cell death, 10(5), 1009-1018 (2005-09-10)
A number of mechanisms have been proposed to explain the etiology of drug-induced lupus (DIL) but the effect of apoptotic and necrotic cell handling has not been previously examined. To evaluate the effect of quinidine and procainamide at therapeutic range
Bin Fan et al.
Journal of neuroimmunology, 244(1-2), 45-50 (2012-01-21)
Meningothelial cells (MECs) form the innermost layer of the meningeal sheath and as such are in direct contact with the cerebrospinal fluid (CSF) likely influencing CSF composition. The CSF space is a site of active immunological processes. To investigate an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service