804622
Bovine Collagen Solution
Type I, 6 mg/mL, ≥95%, sterile filtered, BSE-Free, suitable for biomedical research
Synonym(s):
Collagen I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥95%
form
liquid
concentration
6 mg/mL
shipped in
wet ice
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Highly purified bovine corium pepsin solubilized collagen, atelo collagen, in 0.01 N HCl, pH 2. Predominantly Type I collagen with approximately 5% Type III at 6 mg/ml.
- Sterile filtered.
- BSE-Free.
- ≥95% purity by SDS PAGE.
- Suitable for Biomedical research.
Application
Forms a gel at pH 7 and is ideal for 3D biomaterial scaffolds for cell culture, engineering tissue, and drug delivery.
Legal Information
Product of Collagen Solutions.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Advanced Drug Delivery Reviews, 55, 1631-1649 (2003)
Materials, 3, 1863-1887 (2010)
Angewandte Chemie (International Edition in English), 50, 640-643 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service