Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Collagen Solution
CAS Number:
9007-34-5
Products
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Purity
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
804592
Bovine Collagen Solution,
Type I, 3 mg/mL, ≥95%, sterile filtered, BSE-Free, suitable for biomedical research
Expand
View Pricing
804622
Bovine Collagen Solution,
Type I, 6 mg/mL, ≥95%, sterile filtered, BSE-Free, suitable for biomedical research
Expand
View Pricing
804614
Bovine Collagen Solution,
Type I, Acid soluble telocollagen, 6 mg/mL, sterile filtered, BSE-Free, suitable for biomedical research
Expand
View Pricing