Skip to Content
Merck
All Photos(3)

Key Documents

P11204

Sigma-Aldrich

Perylene

≥99% purity, powder or crystals

Synonym(s):

Dibenz[de,kl]anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1911335
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Perylene, ≥99%

Assay

≥99%

form

powder or crystals

mp

276-279 °C (lit.)

fluorescence

λex 400 nm; λem 445 nm (CH3CN:H2O (8:2))

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

c1cc2cccc3c4cccc5cccc(c(c1)c23)c45

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H

InChI key

CSHWQDPOILHKBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The polycyclic aromatic hydrocarbon perylene is a hydrophobic fluorescent probe. It is highly photostable and displays blue fluorescence. Perylene is used as a dye for spectroscopic studies.

Application

Perylene has been used as a standard along with a mixture of polycyclic aromatic hydrocarbons (PAHs) to determine the spatial distribution and sources of PAHs in surface sediments of rivers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fluorescence probe partitioning between Lo/Ld phases in lipid membranes
Tobias Baumgart
Biochimica et Biophysica Acta, 1768(9), 2182-2194 (2007)
Distribution and sources of polycyclic aromatic hydrocarbons in surface sediments of rivers and an estuary in Shanghai, China
Ying Liu
Environmental Pollution (Barking, Essex : 1987) (2008)
Bernard L. Horecker, Earl R. Stadtman
Current Topics in Cellular Regulation, 2014 null
Bernard Valeur, Mario Nuno Berberan-Santos
Molecular Fluorescence:Principles and Applications (2013)
Andrew Danos et al.
The journal of physical chemistry letters, 6(15), 3061-3066 (2015-08-13)
Photochemical upconversion via triplet-triplet annihilation is a promising technology for improving the efficiency of photovoltaic devices. Previous studies have shown that the efficiency of upconversion depends largely on two rate constants intrinsic to the emitting species. Here, we report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service