795534
Graphene oxide nanocolloids
2 mg/mL, dispersion in H2O
Synonym(s):
NGO, nano-GO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CxHyOz
UNSPSC Code:
12352103
NACRES:
NA.23
Recommended Products
form
dispersion in H2O
Quality Level
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
2 mg/mL
greener alternative category
, Enabling
Related Categories
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". High concentrated graphene oxide sheets provide the prerequisite viscosity to bind the electrode materials together and enable 3D printing. Using water as a green solvent makes this aqueous ink system feasible for processing and drying safety and low cost. Click here for more information.
Application
- Single-layer graphene oxide sheets
- Possible applications:
- Cellular Imaging
- Drug Delivery
- Nanomedicine
- Cancer therapy
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Brixhilda Domi et al.
International journal of molecular sciences, 21(1) (2020-01-02)
The ability of commercial monolayer graphene oxide (GO) and graphene oxide nanocolloids (GOC) to interact with different unicellular systems and biomolecules was studied by analyzing the response of human alveolar carcinoma epithelial cells, the yeast Saccharomyces cerevisiae and the bacteria
Ho Sang DJung et al.
ACS nano, 8(1), 260-268 (2014-01-05)
Melanoma skin cancer is one of the most dangerous skin cancers and the main cause of skin-cancer-related mortality. Hyaluronic acid (HA) has been used as an effective transdermal delivery carrier of chemical drugs and biopharmaceuticals. In this work, a nanographene
Jiayan Luo et al.
Journal of the American Chemical Society, 132(50), 17667-17669 (2010-11-26)
Graphene oxide (GO) nanocolloids-sheets with lateral dimension smaller than 100 nm-were synthesized by chemical exfoliation of graphite nanofibers, in which the graphene planes are coin-stacked along the length of the nanofibers. Since the upper size limit is predetermined by the
Felix Laguna-Teno et al.
Frontiers in microbiology, 11, 1943-1943 (2020-08-28)
Graphene oxide has become a very appealing nanomaterial during the last years for many different applications, but its possible impact in different biological systems remains unclear. Here, an assessment to understand the toxicity of different commercial graphene oxide nanomaterials on
Zhuang Liu et al.
Journal of the American Chemical Society, 130(33), 10876-10877 (2008-07-30)
It is known that many potent, often aromatic drugs are water insoluble, which has hampered their use for disease treatment. In this work, we functionalized nanographene oxide (NGO), a novel graphitic material, with branched polyethylene glycol (PEG) to obtain a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service