Skip to Content
Merck
All Photos(3)

Key Documents

790184

Sigma-Aldrich

Sodium 1-(trifluoromethyl)cyclopropanesulfinate

Synonym(s):

Baran TFCS-Na Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4F3NaO2S
Molecular Weight:
196.12
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

mp

287-292 °C

SMILES string

[O-]S(C1(CC1)C(F)(F)F)=O.[Na+]

InChI

1S/C4H5F3O2S.Na/c5-4(6,7)3(1-2-3)10(8)9;/h1-2H2,(H,8,9);/q;+1/p-1

InChI key

IZLDENBRBYPDNY-UHFFFAOYSA-M

Application

TFCS-Na is a diversification reagent for the C-H Functionalization of several classes of heterocycles. This reagent installs the trifluoromethylcyclopropyl group, which was recently reported to increase metabolic stability compared to compounds containing the tert-butyl group.

Simple Sulfinate Synthesis Enables C-H Trifluoromethylcyclopropanation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Barnes-Seeman et al.
ACS medicinal chemistry letters, 4(6), 514-516 (2014-06-06)
Susceptibility to metabolism is a common issue with the tert-butyl group on compounds of medicinal interest. We demonstrate an approach of removing all the fully sp(3) C-Hs from a tert-butyl group: replacing some C-Hs with C-Fs and increasing the s-character

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service