743232
Sodium triflinate
≥95.0% (T)
Synonym(s):
Langlois reagent, Sodium trifluoromethanesulfinate, Sodium trifluoromethylsulfinate, Trifluoromethanesulfinic acid sodium salt
About This Item
Recommended Products
Quality Level
Assay
≥95.0% (T)
form
powder
reaction suitability
reaction type: C-C Bond Formation
impurities
≤1.0% water (Karl Fischer)
mp
<325 °C
SMILES string
[Na+].[O-]S(=O)C(F)(F)F
InChI
1S/CHF3O2S.Na/c2-1(3,4)7(5)6;/h(H,5,6);/q;+1/p-1
InChI key
KAVUKAXLXGRUCD-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service