Skip to Content
Merck
All Photos(1)

Key Documents

764914

Sigma-Aldrich

Poly(ethylene glycol) 4-cyano-4-(phenylcarbonothioylthio)pentanoate

average Mn 2,000

Synonym(s):

PEG CTA, PEG RAFT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H12NOS2(OCH2CH2)nOCH3
UNSPSC Code:
12352100
NACRES:
NA.23

description

PEG average Mn 2,000 n~44

form

powder

mol wt

average Mn 2,000

mp

47-52 °C

PDI

≤1.1

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization;especially suited for the polymerization of styrene;methacrylate;and methacrylamide monomers to make lithographically and biologically important PEG-block copolymers. Chain Transfer Agent (CTA)
Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service