Skip to Content
Merck
All Photos(1)

Key Documents

760404

Tantalum(V) ethoxide

packaged for use in deposition systems

Synonym(s):

PET, Pentaethoxytantalum, Pentaethyl tantalate, Tantalum pentaethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ta(OC2H5)5
CAS Number:
Molecular Weight:
406.25
Beilstein:
3678999
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.487 (lit.)

bp

155 °C/0.01 mmHg (lit.)

mp

21 °C (lit.)

density

1.566 g/mL at 25 °C (lit.)

SMILES string

CCO[Ta](OCC)(OCC)(OCC)OCC

InChI

1S/5C2H5O.Ta/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5

InChI key

HSXKFDGTKKAEHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tantalum(V) ethoxide precursor is used to deposit ultra thin films of Tantalum oxide and other tantalum containing films by atomic layer deposition and chemical vapor deposition methods

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria-Joao Ribeiro et al.
EJNMMI research, 4, 28-28 (2014-07-10)
Cerebral stroke is a severe and frequent condition that requires rapid and reliable diagnosis. If administered shortly after the first symptoms manifest themselves, IV thrombolysis has been shown to increase the functional prognosis by restoring brain reperfusion. However, a better
Jeffrey D Steinberg et al.
EJNMMI research, 4(1), 15-15 (2014-03-13)
Cerenkov luminescence imaging (CLI) is an emerging imaging technique where visible light emitted from injected beta-emitting radionuclides is detected with an optical imaging device. CLI research has mostly been focused on positive contrast imaging for ascertaining the distribution of the
Kuo-Shyan Lin et al.
Cancer research, 75(2), 387-393 (2014-12-10)
The bradykinin receptor B1R is overexpressed in many human cancers where it might be used as a general target for cancer imaging. In this study, we evaluated the feasibility of using radiolabeled kallidin derivatives to visualize B1R expression in a
Miklós Tóth et al.
PloS one, 10(5), e0125917-e0125917 (2015-05-23)
Molecular imaging of the 18 kD Translocator protein (TSPO) with positron emission tomography (PET) is of great value for studying neuroinflammation in rodents longitudinally. Quantification of the TSPO in rodents is, however, quite challenging. There is no suitable reference region
I Boileau et al.
Molecular psychiatry, 19(12), 1305-1313 (2013-12-11)
Drug addiction has been associated with deficits in mesostriatal dopamine (DA) function, but whether this state extends to behavioral addictions such as pathological gambling (PG) is unclear. Here we used positron emission tomography and the D3 receptor-preferring radioligand [(11)C]-(+)-PHNO during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service