Skip to Content
Merck
All Photos(1)

Key Documents

757918

Sigma-Aldrich

Poly(ethylene glycol) 2-aminoethyl ether acetic acid

average Mn 5,000, cross-linking reagent amine reactive, cross-linking reagent carboxyl reactive, carboxylic acid, amine

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) 2-aminoethyl ether acetic acid, average Mn 5,000

form

solid

Quality Level

mol wt

PEG average Mn 5,000 (n~110)
average Mn 5,000

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reagent type: cross-linking reagent
reactivity: carboxyl reactive

transition temp

Tm 53-58 °C

Ω-end

amine

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Application

  • Bioconjugation
  • Drug Discovery

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service