Skip to Content
Merck
All Photos(2)

Documents

757896

Sigma-Aldrich

Poly(ethylene glycol) 2-aminoethyl ether acetic acid

average Mn 3,500, cross-linking reagent amine reactive, cross-linking reagent carboxyl reactive

Synonym(s):

Polyethylene glycol, Amine-PEG-carboxylic acid, Amino-PEG acetic acid, AminoPEG carboxylic acid, Aminopolyoxyethylene acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC2H4O(C2H4O)nCH2CO2H
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) 2-aminoethyl ether acetic acid, average Mn 3,500

form

solid

mol wt

PEG average Mn 3,400 (n~77)
average Mn 3,500

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

reagent type: cross-linking reagent
reactivity: carboxyl reactive

mp

55-61 °C

Ω-end

amine

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

PEG-PLGA is a biocompatible hydrophobic/hydrophilic drug carrier system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of a theranostic agent: radioiodinated PEGylated PLGA-indocyanine capsules and in vitro determination of their bioaffinity on ovarian, cervical and breast cancer cells.
Akman L, et al.
Journal of radioanalytical and nuclear chemistry, 1-12 (2015)
Synthesis of a theranostic agent: radioiodinated PEGylated PLGA-indocyanine capsules and in vitro determination of their bioaffinity on ovarian, cervical and breast cancer cells.
Akman L, et al.
J. Radioanal. Nucl. Chem., 1-12 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service