Skip to Content
Merck
All Photos(1)

Documents

672130

Sigma-Aldrich

O-(2-Aminoethyl)polyethylene glycol

Mp 5,000

Synonym(s):

Polyethylene glycol, Aminopolyethylene glycol 5,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2CH2O)nH
CAS Number:
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

mol wt

Mp 5,000

Ω-end

hydroxyl

α-end

amine

Application

O-(2-Aminoethyl)polyethylene glycol (PEG-NH2) can be used:
  • As a capping ligand in the synthesis of water-dispersible and non-toxic nanocrystals (ZnS:Mn) for the development of environment friendly materials.
  • In the surface modification of magnetic nanoparticles (MNPs) for a variety of biomedical applications.

Bottomless glass bottle. Contents are inside inserted fused cone.
Reactant for:
  • Preparation of pH responsive self-healing hydrogels formed by boronate-catechol complexation
  • Preparation of core-clickable PEG-branch-azide bivalent-bottle-brush polymers by ROMP
  • Preparation of drug-loaded bivalent-bottle-brush polymers by graft-through ROMP

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential of magnetic nanoparticles for targeted drug delivery
Yang H-W, et al.
Nanotechnology, Science and Applications, 5(4), 73-73 (2012)
Syntheses of Biologically Non-Toxic ZnS: Mn Nanocrystals by Surface Capping with O-(2-aminoethyl) polyethylene Glycol and O-(2-carboxyethyl) polyethylene Glycol Molecules
Kong H-Y, et al.
Bull. Korean Chem. Soc., 34(4), 1181-1187 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service